missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Chlorothionoformate, TRC

Product Code. 30428561
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30428561

Brand: Toronto Research Chemicals TRCP3196505G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenyl Chlorothionoformate
CAS 1005-56-7
Molecular Formula C7H5ClOS
Quantity 5 g
Synonym Carbonochloridothioic Acid O-Phenyl Ester, Chlorothioformic Acid O-Phenyl Ester, NSC 99103, O-Phenyl Carbonochloridothioate, O-Phenyl Chlorothiocarbonate, O-Phenyl Chlorothioformate, O-Phenyl Chlorothionoformate, Phenoxythiocarbonyl Chloride, Phenyl Chlorothiocarbonate, Phenyl Chlorothionoformate, Phenyl Thiochloroformate, Phenyl Thionochloroformate, Phenyl Thioxochloroformate
InChI Formula InChI=1S/C7H5ClOS/c8-7(10)9-6-4-2-1-3-5-6/h1-5H
SMILES ClC(=S)Oc1ccccc1
IUPAC Name O-phenyl chloromethanethioate
Molecular Weight (g/mol) 172.63
Formula Weight 171.975
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.