missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl Chlorothionoformate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Phenyl Chlorothionoformate |
| CAS | 1005-56-7 |
| Molecular Formula | C7H5ClOS |
| Quantity | 1 g |
| Synonym | Carbonochloridothioic Acid O-Phenyl Ester, Chlorothioformic Acid O-Phenyl Ester, NSC 99103, O-Phenyl Carbonochloridothioate, O-Phenyl Chlorothiocarbonate, O-Phenyl Chlorothioformate, O-Phenyl Chlorothionoformate, Phenoxythiocarbonyl Chloride, Phenyl Chlorothiocarbonate, Phenyl Chlorothionoformate, Phenyl Thiochloroformate, Phenyl Thionochloroformate, Phenyl Thioxochloroformate |
| InChI Formula | InChI=1S/C7H5ClOS/c8-7(10)9-6-4-2-1-3-5-6/h1-5H |
| SMILES | ClC(=S)Oc1ccccc1 |
| IUPAC Name | O-phenyl chloromethanethioate |
| Molecular Weight (g/mol) | 172.63 |
| Formula Weight | 171.975 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction