Learn More
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
£27.10 - £127.00
Chemical Identifiers
CAS | 1885-14-9 |
---|---|
Molecular Formula | C7H5ClO2 |
Molecular Weight (g/mol) | 156.57 |
MDL Number | MFCD00000637 |
InChI Key | AHWALFGBDFAJAI-UHFFFAOYSA-N |
Synonym | phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y |
PubChem CID | 15891 |
IUPAC Name | phenyl carbonochloridate |
SMILES | ClC(=O)OC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11444040
|
Thermo Scientific Alfa Aesar
A12618.22 |
100 g |
£27.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450443
|
Thermo Scientific Alfa Aesar
A12618.30 |
250 g |
£47.10
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11458680
|
Thermo Scientific Alfa Aesar
A12618.36 |
500 g |
£86.40
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440443
|
Thermo Scientific Alfa Aesar
A12618.0B |
1000 g | |||||||
Description
Phenyl chloroformate is used in the synthesis of poly(2-(phenoxycarbonyloxy)ethyl methacrylate) and phenyl-(4-vinylphenyl) carbonate. It acts as a precursor of phenyl mixed anhydrides which are used in peptide coupling reactions. It serves as a dehydrating reagent for the conversion of primary amides to nitriles and an intermediate in the synthesis of pharmaceuticals and carbamates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenyl chloroformate is used in the synthesis of poly(2-(phenoxycarbonyloxy)ethyl methacrylate) and phenyl-(4-vinylphenyl) carbonate. It acts as a precursor of phenyl mixed anhydrides which are used in peptide coupling reactions. It serves as a dehydrating reagent for the conversion of primary amides to nitriles and an intermediate in the synthesis of pharmaceuticals and carbamates.
Solubility
Miscible with N,N-dimethylformamide.
Notes
Store in cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases, alcohols and amines. Hydrolyses in water.
Chemical Identifiers
1885-14-9 | |
156.57 | |
AHWALFGBDFAJAI-UHFFFAOYSA-N | |
15891 | |
ClC(=O)OC1=CC=CC=C1 |
C7H5ClO2 | |
MFCD00000637 | |
phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y | |
phenyl carbonochloridate |
Specifications
1885-14-9 | |
1.246 | |
76°C (169°F) | |
1.511 | |
UN2746 | |
Moisture sensitive | |
Miscible with N,N-dimethylformamide. | |
ClC(=O)OC1=CC=CC=C1 | |
156.57 | |
156.57 | |
Phenyl chloroformate |
-38°C | |
188°C to 189°C | |
C7H5ClO2 | |
MFCD00000637 | |
606778 | |
phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y | |
AHWALFGBDFAJAI-UHFFFAOYSA-N | |
phenyl carbonochloridate | |
15891 | |
99% |
Safety and Handling
GHS H Statement
H330-H314-H318-H227-H302-H317
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
May cause an allergic skin reaction.
P210-P235-P260-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H227-H302-H314-H317-H330-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENYL CHLOROFORMATE
missing translation for 'einecsNumber' : 217-547-8
missing translation for 'rtecsNumber' : FG3850000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only