missing translation for 'onlineSavingsMsg'
Learn More

Phenyl bromoacetate, 98%, ACROS Organics™

Catalog Number p-4228868
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
620-72-4
C8H7BrO2
215.046
MFCD00192391
UEWYUCGVQMZMGY-UHFFFAOYSA-N
phenyl bromoacetate, acetic acid, bromo-, phenyl ester, bromoacetic acid phenyl ester, bromoacetic acid, phenyl ester, phenylbromessigester, phenyl bromoacetate #, acmc-209mzb, phenyl 2-bromanylethanoate, ksc352o8b
564919
phenyl 2-bromoacetate
C1=CC=C(C=C1)OC(=O)CBr
This item is not returnable. View return policy
620-72-4
C8H7BrO2
215.046
MFCD00192391
UEWYUCGVQMZMGY-UHFFFAOYSA-N
phenyl bromoacetate, acetic acid, bromo-, phenyl ester, bromoacetic acid phenyl ester, bromoacetic acid, phenyl ester, phenylbromessigester, phenyl bromoacetate #, acmc-209mzb, phenyl 2-bromanylethanoate, ksc352o8b
564919
phenyl 2-bromoacetate
C1=CC=C(C=C1)OC(=O)CBr
Melting Point 26°C to 33°C
Color White to Yellow
Density 1.508g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Formula Weight 215.05
Percent Purity ≥97.5%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Phenyl bromoacetate
Show More Show Less
compliance-icons
Product Identifier
  • Phenyl bromoacetate, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'