missing translation for 'onlineSavingsMsg'
Learn More

Phenyl beta-D-glucopyranoside, 98%

Product Code. 15448429
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448429 25 g 25g
15438429 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448429 Supplier Thermo Scientific Alfa Aesar Supplier No. J66885.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A substrate used for the determination of →-D-glucosidase | CAS: 1464-44-4 | C12H16O6 | 256.25 g/mol

A substrate used for the determination of D-glucosidase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1464-44-4
Molecular Formula C12H16O6
Molecular Weight (g/mol) 256.25
MDL Number MFCD03410292
InChI Key NEZJDVYDSZTRFS-WLEIBRHLNA-N
Synonym phenyl beta-d-glucopyranoside, phenylglucoside, phenyl-beta-d-glucopyranoside, phenol glucoside, phenyl beta-d-glucoside, phenyl-beta-d-glucoside, .beta.-d-glucopyranoside, phenyl, unii-4w3pgi3766, aryl beta-d-glucoside, glucopyranoside, phenyl-, beta-d
PubChem CID 65080
ChEBI CHEBI:28749
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CC=CC=C2)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 173°C to 176°C
Color White
Quantity 25 g
Beilstein 87517
Solubility Information Soluble in water
Optical Rotation −73° (c=1 in Water)
Formula Weight 256.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Phenyl beta-D-glucopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.