missing translation for 'onlineSavingsMsg'
Learn More

Phenyl acetate, 97%

Product Code. p-7026528
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11465137

Brand: Thermo Scientific Alfa Aesar B22561.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-79-2 | C8H8O2 | 136.15 g/mol

Phenyl acetate levels in urine are marker for the diagnosis of some forms of unipolar major depressive disorders. It undergoes Fries rearrangement to form a mixture of o- and p-hydroxyacetophenones which are useful intermediates in manufacture of pharmaceuticals. It is a metabolite of Phenylbutyrate (PB), useful in the treatment of neuroblastoma and lung cancer. An odorant found in strawberries, passion fruit and black tea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (4 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-79-2
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00008699
InChI Key IPBVNPXQWQGGJP-UHFFFAOYSA-N
Synonym acetic acid phenyl ester, phenol acetate, acetylphenol, acetic acid, phenyl ester, acetyl phenol, acetoxybenzene, acetates, acetic acid,phenyl ester, fenylester kyseliny octove, fenylester kyseliny octove czech
PubChem CID 31229
ChEBI CHEBI:8082
IUPAC Name phenyl acetate
SMILES CC(=O)OC1=CC=CC=C1

Specifications

Density 1.075
Boiling Point 195°C to 196°C
Flash Point 76°C (168°F)
Odor Phenol-like
Refractive Index 1.502
Quantity 250 g
Beilstein 636458
Merck Index 14,7267
Solubility Information Soluble in water (4g/L at 20°C).
Formula Weight 136.15
Percent Purity 97%
Chemical Name or Material Phenyl acetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.