missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 4-Hydroxybenzoate, TRC

Product Code. 30133424
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133424 10 g 10g
30010646 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133424 Supplier Toronto Research Chemicals Supplier No. TRCP33533010G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17696-62-7
Molecular Weight (g/mol) 214.22
IUPAC Name phenyl 4-hydroxybenzoate

Specifications

Molecular Formula C13 H10 O3
Quantity 10 g
Synonym Benzoic acid, p-hydroxy-, phenyl ester (7CI,8CI), 4-(Phenoxycarbonyl)phenol, 4-Hydroxybenzoic acid phenyl ester, Phenyl 4-hydroxybenzoate, Phenyl p-hydroxybenzoate, p-Hydroxybenzoic acid phenyl ester, 4-Hydroxybenzoic Acid-Phenyl Ester
InChI Formula InChI=1S/C13H10O3/c14-11-8-6-10(7-9-11)13(15)16-12-4-2-1-3-5-12/h1-9,14H
SMILES Oc1ccc(cc1)C(=O)Oc2ccccc2
Formula Weight 214.063
Chemical Name or Material 4-Hydroxybenzoic Acid-Phenyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.