missing translation for 'onlineSavingsMsg'
Learn More

Phentolamine mesylate, MedChemExpress

Product Code. 16468256
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16468256

Brand: Medchem Express HYB0362A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Phentolamine mesylate (Phentolamine methanesulfonate) is a reversible, non-selective, and orally active blocker of α1 and α2 adrenergic receptor that expands blood vessels to reduce peripheral vascular resistance. Phentolamine mesylate can be used for the research of pheochromocytoma-related hypertension, heart failure and erectile dysfunction.

Phentolamine mesylate (Phentolamine methanesulfonate) is a reversible, non-selective, and orally active blocker of α1 and α2 adrenergic receptor that expands blood vessels to reduce peripheral vascular resistance. Phentolamine mesylate can be used for the research of pheochromocytoma-related hypertension, heart failure and erectile dysfunction.

Specifications

Chemical Name or Material Phentolamine mesylate
CAS 65-28-1
Color White
Purity Grade Notes Research
Molecular Formula C18H23N3O4S
Quantity 100 mg
Synonym Phentolamine methanesulfonate
Solubility Information H2O : ≥ 50 mg/mL (132.46 mM)
SMILES CS(O)(=O)=O.CC1=CC=C(N(C2=CC(O)=CC=C2)CC3=NCCN3)C=C1
Molecular Weight (g/mol) 377.46
Formula Weight 377.46
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.