missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phentolamine mesylate, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Phentolamine mesylate (Phentolamine methanesulfonate) is a reversible, non-selective, and orally active blocker of α1 and α2 adrenergic receptor that expands blood vessels to reduce peripheral vascular resistance. Phentolamine mesylate can be used for the research of pheochromocytoma-related hypertension, heart failure and erectile dysfunction.
Specifications
Specifications
| Chemical Name or Material | Phentolamine mesylate |
| CAS | 65-28-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H23N3O4S |
| Quantity | 500 mg |
| Synonym | Phentolamine methanesulfonate |
| Solubility Information | H2O : ≥ 50 mg/mL (132.46 mM) |
| SMILES | CS(O)(=O)=O.CC1=CC=C(N(C2=CC(O)=CC=C2)CC3=NCCN3)C=C1 |
| Molecular Weight (g/mol) | 377.46 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction