missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Phentolamine Mesylate

Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Phentolamine Mesylate is a non-selective adrenergic α receptor antagonist (pKB = 8.07). Intravenous injection causes hypotension and tachycardia in rats. Intraperitoneal injection in mice increases plasma FGF21 levels.
Specifications
Specifications
| Chemical Name or Material | 3-[[(4,5-Dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]phenol methanesulfonate |
| CAS | 65-28-1 |
| Assay Percent Range | >98% |
| Quantity | 100 mg |
| Molecular Formula | C17H19N3O.CH4O3S |
| Formula Weight | Observed MW: 377.46 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction