missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phentolamine Mesilate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Phentolamine Mesilate
Specifications
Specifications
| Chemical Name or Material | Phentolamine Mesilate |
| CAS | 65-28-1 |
| Analyte or Component Names | Phentolamine Mesilate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H19 N3 O . C H4 O3 S |
| Certifications/Compliance | ISO 17034 |
| Synonym | Phentolamine mesilate, 3-[[(4,5-Dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]phenol methanesulfonate, Phenol, 3-[[(4,5-dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]-, methanesulfonate (1:1), Phenol, 3-[[(4,5-dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]-, monomethanesulfonate (salt) (9CI), Phenol, m-[N-(2-imidazolin-2-ylmethyl)-p-toluidino]-, monomethanesulfonate (salt) (8CI), Phentolamine mesylate, Phentolamine methanesulfonate, Phentolamine methanesulphonate, Regitin methanesulphonate, Regitine mesylate, Regitine methanesulfonate, Rogitine, Vasomax, m-[N-(2-Imidazolin-2-ylmethyl)-p-toluidino]phenol methanesulfonate |
| InChI Formula | InChI=1S/C17H19N3O.CH4O3S/c1-13-5-7-14(8-6-13)20(12-17-18-9-10-19-17)15-3-2-4-16(21)11-15;1-5(2,3)4/h2-8,11,21H,9-10,12H2,1H3,(H,18,19);1H3,(H,2,3,4) |
| SMILES | Cc1ccc(cc1)N(CC2=NCCN2)c3cccc(O)c3.CS(=O)(=O)O |
| IUPAC Name | 3-[N-(4,5-dihydro-1H-imidazol-2-ylmethyl)-4-methylanilino]phenol;methanesulfonic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction