missing translation for 'onlineSavingsMsg'
Learn More

Phenprocoumon, MedChemExpress

Product Code. p-200101731
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16494187

Brand: Medchem Express HYA0145

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Phenprocoumon is a coumarin derivative that acts as a long acting oral anticoagulant and an antagonist of vitamin K.

Phenprocoumon is a coumarin derivative that acts as a long acting oral anticoagulant and an antagonist of vitamin K.

Specifications

Chemical Name or Material Phenprocoumon
CAS 435-97-2
Color White
Purity Grade Notes Research
Molecular Formula C18H16O3
Quantity 50 mg
Solubility Information DMSO : ≥ 125 mg/mL (445.92 mM)
SMILES O=C1C(C(C2=CC=CC=C2)CC)=C(O)C3=CC=CC=C3O1
Molecular Weight (g/mol) 280.32
Formula Weight 280.32
Percent Purity 98.09%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.