missing translation for 'onlineSavingsMsg'
Learn More

Phenoxybenzamine hydrochloride

Product Code. 15454389
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454389 250 mg 250mg
15444389 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454389 Supplier Thermo Scientific Alfa Aesar Supplier No. J61900.MD
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63-92-3 | C18H23Cl2NO | 340.288 g/mol

Selective alpha adrenergic blocking agent. Irreversible calmodulin inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Selective alpha adrenergic blocking agent
  • Irreversible calmodulin inhibitor
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-92-3
Molecular Formula C18H23Cl2NO
Molecular Weight (g/mol) 340.288
MDL Number MFCD00055152
InChI Key VBCPVIWPDJVHAN-UHFFFAOYSA-N
Synonym phenoxybenzamine hydrochloride, dibenzyline, phenoxybenzamine hcl, n-benzyl-n-2-chloroethyl-1-phenoxypropan-2-amine hydrochloride, dibenzyline hydrochloride, phenoxybenzamine.hcl, bensylyt nen, phenoxybenzamine chloride, benzene methanamine
PubChem CID 5284441
ChEBI CHEBI:8078
IUPAC Name N-benzyl-N-(2-chloroethyl)-1-phenoxypropan-2-amine;hydrochloride
SMILES CC(COC1=CC=CC=C1)N(CCCl)CC2=CC=CC=C2.Cl

Specifications

Color White
Quantity 250 mg
Merck Index 14,7256
Formula Weight 340.29
Physical Form Powder
Chemical Name or Material Phenoxybenzamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.