Learn More
Phenoxybenzamine hydrochloride, Thermo Scientific™
Brand:  Thermo Scientific Alfa Aesar J61900.03
 View more versions of this product
Product Code. 15444389
  • ONLINE EXCLUSIVE PRICE
    £37.40
    £36.60 / 1g
    Save £0.80
    2% Off
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Selective alpha adrenergic blocking agent
  • Irreversible calmodulin inhibitor
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS Documents