missing translation for 'onlineSavingsMsg'
Learn More

Phenoxybenzamine hydrochloride, MedChemExpress

Product Code. 16422136
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
1 g
Unit Size:
1g
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422136 500 mg 500mg
16461416 200 mg 200mg
16452169 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16422136 Supplier Medchem Express Supplier No. HYB0431A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Phenoxybenzamine hydrochloride is a nonselective, irreversible, orally active α-adrenoceptor antagonist that is commonly used for the research of hypertension, specifically caused by pheochromocytoma. Phenoxybenzamine hydrochloride also shows antitumor activity.

Phenoxybenzamine hydrochloride is a nonselective, irreversible, orally active α-adrenoceptor antagonist that is commonly used for the research of hypertension, specifically caused by pheochromocytoma. Phenoxybenzamine hydrochloride also shows antitumor activity.

Specifications

Chemical Name or Material Phenoxybenzamine hydrochloride
CAS 63-92-3
Color White
Purity Grade Notes Research
Molecular Formula C18H23Cl2NO
Quantity 500 mg
Solubility Information DMSO : 100 mg/mL (293.87 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.97 mM; Need ultrasonic)
SMILES CC(COC1=CC=CC=C1)N(CCCl)CC2=CC=CC=C2.Cl
Molecular Weight (g/mol) 340.29
Formula Weight 340.29
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.