missing translation for 'onlineSavingsMsg'
Learn More

Phenoxyacetonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11434014
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434014 5 g 5g
11444014 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434014 Supplier Thermo Scientific Alfa Aesar Supplier No. A11981.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3598-14-9
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00052017
InChI Key VLLSCJFPVSQXDM-UHFFFAOYSA-N
Synonym phenoxyacetonitrile, acetonitrile, phenoxy, 2-phenoxyethanenitrile, phenoxy-acetonitrile, 2-phenoxy-acetonitrile, acmc-1ajch, acetonitrile,2-phenoxy
PubChem CID 241641
IUPAC Name 2-phenoxyacetonitrile
SMILES N#CCOC1=CC=CC=C1

Specifications

Density 1.09
Boiling Point 76°C to 78°C (0.05 mmHg)
Refractive Index 1.525
Quantity 5 g
UN Number UN3276
Beilstein 2206192
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material Phenoxyacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.