missing translation for 'onlineSavingsMsg'
Learn More

Phenoxy-d5-acetic Acid, TRC

Product Code. 30423144
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30423144 5 mg
30408990 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30423144 Supplier Toronto Research Chemicals Supplier No. TRCP3181025MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154492-74-7
Molecular Formula C8 2H5 H3 O3
Molecular Weight (g/mol) 157.18
Synonym Acetic acid, phenoxy-d5- (9CI), Phenoxy-d5-acetic acid, Phenoxyacetic acid D5 (phenoxy-D5), 2-Phenoxy-d5-acetic acid, 2-(Phenyl-d5-oxy)acetic acid, Glycolic acid phenyl-d5 ether, Phenoxyacetic acid-d5, Pentadeuteriophenoxyethanoic acid
IUPAC Name 2-(2,3,4,5,6-pentadeuteriophenoxy)acetic acid
SMILES [2H]c1c([2H])c([2H])c(OCC(=O)O)c([2H])c1[2H]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C8H8O3/c9-8(10)6-11-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)/i1D,2D,3D,4D,5D
Formula Weight 157.0787
Chemical Name or Material Phenoxy-d5-acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.