missing translation for 'onlineSavingsMsg'
Learn More

Phenothiazine, TRC

Product Code. 30135153
Change view
Click to view available options
Quantity:
50 mg
25 g
500 g
Unit Size:
25g
500g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135153 500 g 500g
30003536 25 g 25g
17903535 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135153 Supplier Toronto Research Chemicals Supplier No. TRCP318040500G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-84-2
Molecular Formula C12 H9 N S
Molecular Weight (g/mol) 199.27
Synonym Phenothiazine, Promethazine Hydrochloride Imp. A (EP), 10H-Phenothiazine, Antage TDP, Contaverm, Danikoropa, Dibenzo-1,4-thiazine, Dibenzothiazine, ENT 38, Early bird wormer, Feeno, Fenoverm, NSC 2037, Nemazene, Nexarbol, Orimon, Padophene, Penthazine, Phenegic, Phenoverm, Phenovis, Phenoxur, Phenthiazine, Phenzeen, Reconox, Thiodiphenylamine
IUPAC Name 10H-phenothiazine
SMILES N1c2ccccc2Sc3ccccc13

Specifications

Quantity 500 g
InChI Formula InChI=1S/C12H9NS/c1-3-7-11-9(5-1)13-10-6-2-4-8-12(10)14-11/h1-8,13H
Formula Weight 199.0456
Percent Purity >95%
Chemical Name or Material Phenothiazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.