missing translation for 'onlineSavingsMsg'
Learn More

Phenothiazine S,S-Dioxide, TRC

Product Code. 30137735
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137735 500 mg 500mg
30132971 1 g 1g
30008842 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137735 Supplier Toronto Research Chemicals Supplier No. TRCP297600500MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1209-66-1
Molecular Formula C12H9NO2S
Molecular Weight (g/mol) 231.27
Synonym 5,5-Dioxophenothiazine;
IUPAC Name 10H-phenothiazine 5,5-dioxide
SMILES O=S1(=O)c2ccccc2Nc3ccccc13

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H9NO2S/c14-16(15)11-7-3-1-5-9(11)13-10-6-2-4-8-12(10)16/h1-8,13H
Formula Weight 231.0354
Percent Purity >95%
Chemical Name or Material Phenothiazine S,S-Dioxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.