missing translation for 'onlineSavingsMsg'
Learn More

Phenothiazine, Mikromol™

Product Code. 17911226
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17911226 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17911226 Supplier Mikromol Supplier No. MM0157.02
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Phenothiazine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenothiazine
CAS 92-84-2
Analyte or Component Names Phenothiazine
Shipping Condition Room Temperature
Molecular Formula C12 H9 N S
Certifications/Compliance ISO 17025
Synonym Promethazine Hydrochloride Imp. A (EP), Promethazine Imp. A (EP), Phenothiazine, 10H-Phenothiazine, Antage TDP, Contaverm, Danikoropa, Dibenzo-1,4-thiazine, Dibenzothiazine, ENT 38, Early bird wormer, Feeno, Fenoverm, NSC 2037, Nemazene, Nexarbol, Orimon, Padophene, Penthazine, Phenegic, Phenoverm, Phenovis, Phenoxur, Phenthiazine, Phenzeen, Reconox, Thiodiphenylamine, Promethazine Hydrochloride Impurity A, Promethazine Impurity A
InChI Formula InChI=1S/C12H9NS/c1-3-7-11-9(5-1)13-10-6-2-4-8-12(10)14-11/h1-8,13H
SMILES N1c2ccccc2Sc3ccccc13
IUPAC Name 10H-phenothiazine
Molecular Weight (g/mol) 199.27
Quantity 100 mg
Formula Weight 199.0456 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.