missing translation for 'onlineSavingsMsg'
Learn More

Hach Pacific Pty Ltd Phenolphthalein Indicator Powder Pillows

Product Code. 10549783
Click to view available options
Quantity:
100/Pk.
This item is not returnable. View return policy

Product Code. 10549783

Brand: Hach Pacific Pty Ltd 94299

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

pH indicator for use in titrations.

pH indicator for use in titrations. Color change: colorless to pink at ∼pH 8. 3. Pack of 100 Powder Pillows. pH indicator for use in titrations. Color change: colorless to pink at ∼pH 8. 3. Pack of 100 Powder Pillows.

Specifications
Method: 8219 (MO)
Method Number 2: 8010 (P)
Method Number 3: 8201 (MO)
Method Number 4: 8202 (P)
Number of tests: 100
Platform: Powder Pillows
Quantity: 100 /pk
Range: Varies by method

Specifications

Quantity 100/Pk.
Range Varies by method
Type Water Testing Kit Supplies and Reagents
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.