missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenolphthalein, 2% soln. in 95% ethanol

Product Code. 15444299
Change view
Click to view available options
Quantity:
100 mL
1000 mL
Unit Size:
1000mL
100mL
Product Code. Quantity unitSize
15444299 100 mL 100mL
15454299 1000 mL 1000mL
2 options
This item is not returnable. View return policy

Product Code. 15444299

Brand: Thermo Scientific Chemicals J61785.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

pH indicator | CAS: 77-09-8 | C20H14O4 | 318.33 g/mol

It is used as a laboratory reagent and pH indicator. Phenolphthalein is a common indicator to use in neutralization titrations. Phenolphthalein exerts laxative effects by stimulating the intestinal mucosa and constricting smooth muscles. It is used medicinally as a cathartic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
Synonym phenolphthalein, phthalimetten, euchessina, phthalin, espotabs, phenolax, purgophen, koprol, laxogen, trilax
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Color Colorless
UN Number UN1987
Concentration 2% soln. in 95% ethanol
Merck Index 14,7243
Solubility Information Miscible in water.
Formula Weight 318.33
Percent Purity 95% (ethanol)
Odor Alcohol-like
Quantity 100 mL
Flash Point 17°C (63°F)
Physical Form Liquid
Chemical Name or Material Phenolphthalein
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.