missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein (0.04% Solution in Ethanol), TRC
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
Specifications
Specifications
| Chemical Name or Material | Phenolphthalein |
| CAS | 77-09-8 |
| Molecular Formula | C20 H14 O4 |
| Quantity | 50 mL |
| Synonym | Phenolphthalein, 3,3-bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one, 1(3H)-Isobenzofuranone, 3,3-bis(4-hydroxyphenyl)-, Phenolphthalein (8CI), 3,3-Bis(4-hydroxyphenyl)-1(3H)-isobenzofuranone, 3,3-Bis(4-hydroxyphenyl)phthalide, 3,3-Bis(p-hydroxyphenyl)phthalide, Euchessina, Koprol, Laxogen, Lilo, NSC 10464, NSC 215214, Phthalimetten, Phthalin, Purga, Purgen, Purgophen, Spulmako-lax, TY 9, Trilax |
| InChI Formula | InChI=1S/C20H14O4/c21-15-9-5-13(6-10-15)20(14-7-11-16(22)12-8-14)18-4-2-1-3-17(18)19(23)24-20/h1-12,21-22H |
| SMILES | Oc1ccc(cc1)C2(OC(=O)c3ccccc23)c4ccc(O)cc4 |
| IUPAC Name | 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one |
| Molecular Weight (g/mol) | 318.32 |
| Formula Weight | 318.0892 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction