missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Phenolic Screw Threaded Caps

Product Code. 10383563 Shop All Corning Products
Click to view available options
Height (Metric):
12.5mm
13.5mm
Unit Size:
Pack of 12
Pack of 288
This item is not returnable. View return policy

Product Code. 10383563

Brand: Corning™ 999924

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.

These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.

TRUSTED_SUSTAINABILITY

Specifications

Closure Size 24-400
Liner Material Rubber
Product Type Screw Cap
Autoclavable Autoclavable
Material Phenolic
Color Black
Height (Metric) 12.5mm
No. per Pack 12 Pack
No. per Case 12 Pack
Packaging 12 Pack
Quantity 12 Pack
For Use With (Equipment) Culture Tubes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.