missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red Stain, MP Biomedicals

Product Code. 11404815 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL

Product Code. 11404815

Brand: MP Biomedicals 091690049

Please to purchase this item. Need a web account? Register with us today!

0.5% (w/v) Solution Storage temperature: 15-30°C

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenol Red Stain
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.