missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red Sodium Salt, TRC

Product Code. 30419166
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419166 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419166 Supplier Toronto Research Chemicals Supplier No. TRCP2953155G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenol Red Sodium Salt
CAS 34487-61-1
Molecular Formula C19H14O5S . Na
Quantity 5 g
Synonym 3H-2,1-Benzoxathiole','Phenol deriv., Phenol Red Sodium, Phenol Red','Sodium Salt, Phenolsulfonephthalein Sodium, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis-phenol S,S-Dioxide Monosodium Salt
InChI Formula InChI=1S/C19H14O5S.Na/c20-15-9-5-13(6-10-15)19(14-7-11-16(21)12-8-14)17-3-1-2-4-18(17)25(22,23)24-19;/h1-12,20-21H;/q;+1/p-1
SMILES [Na+].Oc1ccc(cc1)C2(OS(=O)(=O)c3ccccc23)c4ccc([O-])cc4
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1λ^{6}-benzoxathiol-3-yl]phenolate
Molecular Weight (g/mol) 376.4
Formula Weight 376.0381
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.