Learn More
Thermo Scientific Chemicals Phenol Red sodium salt, ACS
pH indicator
£22.10 - £59.00
Chemical Identifiers
CAS | 34487-61-1 |
---|---|
Molecular Formula | C19H13NaO5S |
Molecular Weight (g/mol) | 376.358 |
MDL Number | MFCD00066901 |
InChI Key | HKHYOKBQJILTEI-UHFFFAOYSA-M |
PubChem CID | 23686673 |
IUPAC Name | sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate |
SMILES | C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387148
|
Thermo Scientific Chemicals
015987.06 |
5 g |
£22.10
5g |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
11377148
|
Thermo Scientific Chemicals
015987.14 |
25 g |
£59.00
25g |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
Description
Phenol red sodium salt is a pH indicator commonly used in cell culture for monitoring pH changes. It is used in the pH ranges between 6.8 and 8.2. It is involved in the studies of gastrointestinal and other systems. It can be used to check hydrogen peroxide in supernatants from cultured macrophages in multiwell plates. Further, it is used as a diagnostic aid for determination of renal function.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
34487-61-1 | |
376.358 | |
HKHYOKBQJILTEI-UHFFFAOYSA-M | |
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate |
C19H13NaO5S | |
MFCD00066901 | |
23686673 | |
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+] |
Specifications
285°C (decomposition) | |
C19H13NaO5S | |
6260026 | |
Soluble in water,alcohol,aqueous alkali hydroxides and carbonates. | |
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+] | |
376.358 | |
376.37 | |
Crystalline |
34487-61-1 | |
MFCD00066901 | |
14,7247 | |
HKHYOKBQJILTEI-UHFFFAOYSA-M | |
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate | |
23686673 | |
ACS | |
Phenol Red sodium salt |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 252-057-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only