missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol Red sodium salt, ACS

Product Code. 11377148
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377148 25 g 25g
11387148 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377148 Supplier Thermo Scientific Chemicals Supplier No. 015987.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

pH indicator | CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol

Phenol red sodium salt is a pH indicator commonly used in cell culture for monitoring pH changes. It is used in the pH ranges between 6.8 and 8.2. It is involved in the studies of gastrointestinal and other systems. It can be used to check hydrogen peroxide in supernatants from cultured macrophages in multiwell plates. Further, it is used as a diagnostic aid for determination of renal function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
MDL Number MFCD00066901
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

Melting Point 285°C (decomposition)
Beilstein 6260026
Merck Index 14,7247
Solubility Information Soluble in water,alcohol,aqueous alkali hydroxides and carbonates.
Formula Weight 376.37
Grade ACS
Quantity 25 g
Physical Form Crystalline
Chemical Name or Material Phenol Red sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.