missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Phenol Red sodium salt
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Product Type | Dye Reagents |
| Quantity | 5 g |
| Shipping Condition | Room temperature |
| Synonym | Phenolsulfonephthalein sodium salt |
| CAS | 34487-61-1 |
| Molecular Weight (g/mol) | 376.36 g/mol |
| Solubility | DMSO : 5 mg/mL (13.29 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble) |
| Target | HBV |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?