missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Phenol Red sodium salt

Product Code. p-200104498
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
250 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463816 250 g 25g
16411168 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16463816 Supplier MedChemExpress Supplier No. HYD0169A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Phenol red (Phenolsulfonephthalein) sodium salt is a pH indicator dye, relying on a distinct color change from pink to yellow in case of a positive reaction .

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Product Type Dye Reagents
Quantity 250 g
Shipping Condition Room temperature
Synonym Phenolsulfonephthalein sodium salt
CAS 34487-61-1
Molecular Weight (g/mol) 376.36 g/mol
Solubility DMSO : 5 mg/mL (13.29 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble)
Target HBV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.