missing translation for 'onlineSavingsMsg'
Learn More

Phenol, MP Biomedicals™

Product Code. 11444966 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444966 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11444966 Supplier MP Biomedicals Supplier No. 04818048
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phenol, Ultra Pure Grade, White Solid

  • Phenol is useful for extraction of viral, plasmid and phage DNAs, and to solubilize and dissociate proteins.
  • It is used as a general disinfectant, either in solution or mixed with slaked lime, etc., for toilets, stables, cesspools, floors, drains, etc.; for the manufacture of colorless or light-colored artificial resins, many medical and industrial organic compounds and dyes; as a reagent in chemical analysis.
  • Aqueous solutions are used as topical anesthetic; topical antiseptic; topical antipruritic. Phenol is used in DNA extractions for removing protein.

Specifications

CAS 108-95-2
Melting Point 182°C (literature)
Color White
Density 1.071g/mL at 25°C (Lit.)
Boiling Point 182°C (Literature)
Flash Point 174.2°F/79°C (Lit.)
Molecular Formula C6H6O
MDL Number MFCD00002143
Quantity 500 g
UN Number r
Beilstein 969616
Residue after Evaporation <0.05%
Synonym Hydroxybenzene, Carbolic acid, Phenic acid, Phenylic acid
Solubility Information Soluble in aqueous solution (5% - clear, colorless solution).
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES OC1=CC=CC=C1
IUPAC Name phenol
Molecular Weight (g/mol) 94.11
Vapor Density 3.24 (vs air)(Lit.)
Vapor Pressure 0.09 psi (55°C); 0.36 mmHg (20°C)(Lit.)
Percent Purity 0.999
Physical Form Solid
Assay Ultra Puregrade
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.