missing translation for 'onlineSavingsMsg'
Learn More

Phenol-Indo-2,6-Dichlorophenol Sodium Salt, Pure, Tablets, Fisher Chemical

Product Code. 10040300 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
20 Tablets
Packaging:
Amber glass bottle
Unit Size:
20 tablets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10040300 20 Tablets Amber glass bottle 20 tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10040300 Supplier Fisher Chemical Supplier No. P/2392/82
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 620-45-1
Color Green
Physical Form Powder/Solid
Quantity 20 Tablets
Molecular Formula C12H6Cl2NNaO2
MDL Number MFCD00150014
Synonym 2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt
InChI Key FHLDWQLHDYCXKI-UHFFFAOYSA-M
SMILES [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1
IUPAC Name sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
Molecular Weight (g/mol) 290.07
PubChem CID 23697355
ChEBI CHEBI:948
Formula Weight 325.9701g/mol
Packaging Amber glass bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.