missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Acidity | Complying |
| Flash Point | 79 °C |
| Packaging | Plastic Bottle |
| Quantity | 500 g |
| Percent Purity | 99.5 to 100.5% |
| Grade | Reagent Pharmacopeia Organics |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction