missing translation for 'onlineSavingsMsg'
Learn More

Phenol Detached Crystals, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10734381 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 kg
Unit Size:
1kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10734381 500 g 500g
10569380 1 kg 1kg
10345810 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10734381 Supplier Fisher Chemical Supplier No. P/2360/53
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

C6H6O, CAS Number-108-95-2, oxybenzene, hydroxybenzene, carbolic acid, phenyl hydroxide, monophenol, phenyl hydrate, phenic acid, phenylic acid, phenylic alcohol, benzenol, 1kg, 182 deg.C, CHEBI:15882, Colorless to White, 94.11g/mol, ISWSIDIOOBJBQZ-UHFFFAOYSA-N, 39 deg.C, 2143, 94.113 | CAS: 108-95-2 | C6H6O | 94.11 g/mol

C6H6O, CAS Number-108-95-2, oxybenzene, hydroxybenzene, carbolic acid, phenyl hydroxide, monophenol, phenyl hydrate, phenic acid, phenylic acid, phenylic alcohol, benzenol, 1kg, 182 deg.C, CHEBI:15882, Colorless to White, 94.11g/mol, ISWSIDIOOBJBQZ-UHFFFAOYSA-N, 39 deg.C, 2143, 94.113

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Melting Point 39°C
Color Colorless to White
pH 6
Boiling Point 182°C
Packaging Plastic powder jar
Quantity 500 g
Viscosity 3.437 mPaS (50°C)
Formula Weight 94.11g/mol
Vapor Pressure 0.4mbar at 20°C
Physical Form Crystalline Solid
Chemical Name or Material Phenol Detached Crystals
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.