missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol (Crystallized), Fisher BioReagents
Click to view available options
Amber Glass/Poison Pack; 100g
Description
Phenol is used in nucleic acid purification procedures.
Specifications
CAS | 108-95-2 |
Melting Point | 39°C |
Color | Colorless |
Density | 1.070g/cm³ |
pH | 4 to 5 |
Boiling Point | 182°C |
Assay Percent Range | ≥99.0 % |
Packaging | Amber Glass/Poison Pack |
Molecular Formula | C6H6O |
Linear Formula | C6H5OH |
Show More |
Safety and Handling
Product Identifier
- Phenol
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- H314-Causes severe skin burns and eye damage.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'