missing translation for 'onlineSavingsMsg'
Learn More

Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.), Fisher BioReagents™

Product Code. 10699543
Change view
Click to view available options
Quantity:
100 mL
400 mL
Packaging:
Coated Amber Glass/Poison Pack
Unit Size:
100mL
400mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10699543 400 mL Coated Amber Glass/Poison Pack 400mL
10205113 100 mL Coated Amber Glass/Poison Pack 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10699543 Supplier Fisher BioReagents Supplier No. BP1754I400

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.)

This mixture is used to purify total RNA from mixtures containing RNA, DNA, and protein.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Merck Index 15, 7354
Percent Purity ≥99%
Identification Pass Test
Packaging Coated Amber Glass/Poison Pack
pH 4.3
Quantity 400 mL
Physical Form Liquid
Chemical Name or Material Phenol/Chloroform/Isoamyl Alcohol, 125:24:1 Mixture
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenol/Chloroform/Isoamyl Alcohol, pH 4.3
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental information
  • MIXTURE LIST-Contain: Phenol, Chloroform,Isoamyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.