missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phenobarbital Monoclonal Antibody (PB9-30.17.4.16)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA110741
This item is not returnable.
View return policy
Description
MA1-10741 detects Phenobarbital. MA1-10741 has been successfully used in ELISA applications. The MA1-10741 immunogen is Phenobarbital, conjugated to KLH.
Phenobarbital is a long-acting barbituric acid derivative with antipsychotic property. Phenobarbital binds to and activates the gamma-aminobutyric acid (GABA)-A receptor, thereby mimicking the inhibitory actions of GABA in the brain. The activation effects of the phenobarbital-receptor-ionophore complex include increased frequency of chloride channel openings, membrane hyperpolarization and ultimately synaptic inhibition and decreased neuronal excitability. In addition, this agent inhibits glutamate induced depolarization. [National Center for Biotechnology Information. PubChem Compound Database; CID=4763].
Specifications
| Phenobarbital | |
| Monoclonal | |
| 1.7 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 |
| ELISA | |
| PB9-30.17.4.16 | |
| Unconjugated | |
| C12H12N2O3; Luminal; Phenobarbitol; Phenobarbitone; Phenylethylbarbiturate | |
| Phenobarbital, conjugated to KLH. | |
| 500 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction