missing translation for 'onlineSavingsMsg'
Learn More

Phenindione, MedChemExpress

Product Code. 16484327
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484327 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16484327 Supplier Medchem Express Supplier No. HYB0325
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Phenindione is an anticoagulant which functions as a Vitamin K antagonist.

Phenindione is an anticoagulant which functions as a Vitamin K antagonist.

Specifications

Chemical Name or Material Phenindione
CAS 83-12-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H10O2
Quantity 50 mg
Synonym Rectadione
Solubility Information DMSO : ≥ 100 mg/mL (449.96 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C(C2=CC=CC=C2)C(C3=C1C=CC=C3)=O
Molecular Weight (g/mol) 222.24
Formula Weight 222.24
Percent Purity 99.22%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.