missing translation for 'onlineSavingsMsg'
Learn More

Phenetole, 98+%

Product Code. 11422897
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422897 2500 mL 2500mL
11402897 100 mL 100mL
11412897 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422897 Supplier Thermo Scientific Alfa Aesar Supplier No. A15501.0F
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 103-73-1 | C8H10O | 122.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-73-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00009090
InChI Key DLRJIFUOBPOJNS-UHFFFAOYSA-N
Synonym phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f
PubChem CID 7674
ChEBI CHEBI:67129
IUPAC Name ethoxybenzene
SMILES CCOC1=CC=CC=C1

Specifications

Melting Point -30°C
Density 0.967
Boiling Point 169°C to 170°C
Flash Point 57°C (134°F)
Odor Aromatic
Refractive Index 1.507
Quantity 2500 mL
UN Number UN3271
Beilstein 636270
Merck Index 14,7229
Formula Weight 122.17
Percent Purity ≥98%
Chemical Name or Material Phenetole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.