missing translation for 'onlineSavingsMsg'
Learn More

Phenethyl Isocyanate, TRC

Product Code. 30424839
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424839 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424839 Supplier Toronto Research Chemicals Supplier No. TRCP33534010G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenethyl Isocyanate
CAS 1943-82-4
Molecular Formula C9H9NO
Quantity 10 g
Synonym (2-Isocyanatoethyl)benzene, 2-Phenylethyl Isocyanate, Phenethyl Isocyanate, beta-Phenylethyl Isocyanate, Phenethyl Alcohol','Isocyanate, Isocyanic Acid','Phenethyl Ester
InChI Formula InChI=1S/C9H9NO/c11-8-10-7-6-9-4-2-1-3-5-9/h1-5H,6-7H2
SMILES O=C=NCCc1ccccc1
IUPAC Name 2-isocyanatoethylbenzene
Molecular Weight (g/mol) 147.17
Formula Weight 147.0684
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.