missing translation for 'onlineSavingsMsg'
Learn More

Phenethyl isocyanate, 99%

Product Code. 10117972 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10117972 25 mL Glass bottle 25mL
10227982 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10117972 Supplier Thermo Scientific Acros Supplier No. 380660250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1943-82-4 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1943-82-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00037044
InChI Key HACRKYQRZABURO-UHFFFAOYSA-N
Synonym phenethyl isocyanate, 2-isocyanatoethyl benzene, phenylethyl isocyanate, 2-phenylethyl isocyanate, 2-phenylethylisocyanate, phenylethylisocyanate, phenethylisocyanate, benzene, 2-isocyanatoethyl, 2-isocyanato-ethyl-benzene, 2-phenethyl isocyanate
PubChem CID 160602
ChEBI CHEBI:60067
IUPAC Name 2-isocyanatoethylbenzene
SMILES C1=CC=C(C=C1)CCN=C=O

Specifications

Color Colorless to Yellow
Density 1.0630g/mL
Boiling Point 210°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5(CH2)2NCO
Refractive Index 1.5210 to 1.523
Quantity 25 mL
Specific Gravity 1.063
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in toluene
Formula Weight 147.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Phenethyl isocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenethyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.