missing translation for 'onlineSavingsMsg'
Learn More

Phenethyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog Number p-3213038
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
1943-82-4
C9H9NO
147.18
MFCD00037044
HACRKYQRZABURO-UHFFFAOYSA-N
phenethyl isocyanate, 2-isocyanatoethyl benzene, phenylethyl isocyanate, 2-phenylethyl isocyanate, 2-phenylethylisocyanate, phenylethylisocyanate, phenethylisocyanate, benzene, 2-isocyanatoethyl, 2-isocyanato-ethyl-benzene, 2-phenethyl isocyanate
160602
CHEBI:60067
2-isocyanatoethylbenzene
C1=CC=C(C=C1)CCN=C=O
This item is not returnable. View return policy
1943-82-4
C9H9NO
147.18
MFCD00037044
HACRKYQRZABURO-UHFFFAOYSA-N
phenethyl isocyanate, 2-isocyanatoethyl benzene, phenylethyl isocyanate, 2-phenylethyl isocyanate, 2-phenylethylisocyanate, phenylethylisocyanate, phenethylisocyanate, benzene, 2-isocyanatoethyl, 2-isocyanato-ethyl-benzene, 2-phenethyl isocyanate
160602
CHEBI:60067
2-isocyanatoethylbenzene
C1=CC=C(C=C1)CCN=C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.0630g/mL
Boiling Point 210°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5(CH2)2NCO
Refractive Index 1.5210 to 1.523
Quantity 5 mL
Specific Gravity 1.063
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in toluene
Formula Weight 147.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Phenethyl isocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenethyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'