missing translation for 'onlineSavingsMsg'
Learn More

Phenethyl alcohol, 99%

Product Code. 10792754 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic Drum
Unit Size:
1 L
10 L
100mL
2.5 L
250mL
5mL
This item is not returnable. View return policy

Product Code. 10792754

Brand: Thermo Scientific Acros 130180100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60-12-8 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-12-8
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002886
InChI Key WRMNZCZEMHIOCP-UHFFFAOYSA-N
Synonym phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
PubChem CID 6054
ChEBI CHEBI:49000
SMILES OCCC1=CC=CC=C1

Specifications

Melting Point -27.0°C
Color Colorless
Density 1.0200g/mL
Boiling Point 219.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic Drum
Linear Formula C6H5CH2CH2OH
Refractive Index 1.5300 to 1.5350
Quantity 10 L
Beilstein 06, 478
Merck Index 15, 7338
Specific Gravity 1.02
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: miscible with alcohol and ether
IUPAC Name 2-phenylethanol
Viscosity 7.58 mPa.s (25°C)
Formula Weight 122.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Phenethyl alcohol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.