missing translation for 'onlineSavingsMsg'
Learn More

Phenethyl alcohol, 99%

Product Code. 10123230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic Drum
Unit Size:
1 L
10 L
100mL
2.5 L
250mL
5mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10123230 1 L Glass bottle 1 L
10133230 5 mL Glass bottle 5mL
10246563 100 mL Glass bottle 100mL
10521784 250 mL Glass bottle 250mL
10562141 2.5 L Glass bottle 2.5 L
10792754 10 L Plastic Drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10123230 Supplier Thermo Scientific Acros Supplier No. 130180010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60-12-8 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-12-8
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002886
InChI Key WRMNZCZEMHIOCP-UHFFFAOYSA-N
Synonym phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
PubChem CID 6054
ChEBI CHEBI:49000
SMILES OCCC1=CC=CC=C1

Specifications

Melting Point -27.0°C
Color Colorless
Density 1.0200g/mL
Boiling Point 219.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2CH2OH
Refractive Index 1.5300 to 1.5350
Quantity 1 L
Beilstein 06, 478
Merck Index 15, 7338
Specific Gravity 1.02
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: miscible with alcohol and ether
IUPAC Name 2-phenylethanol
Viscosity 7.58 mPa.s (25°C)
Formula Weight 122.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Phenethyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • Phenethyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Phenethyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.