missing translation for 'onlineSavingsMsg'
Learn More

Phenazopyridine hydrochloride, 99%, analytical standard

Product Code. 11936421 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 11936421

Brand: Thermo Scientific Acros 441921000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 136-40-3 | C11H11N5·ClH | 249.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 136-40-3
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5 to max. 101.5% min
Molecular Formula C11H11N5·ClH
Quantity 100 mg
Merck Index 15,7324
Synonym phenazopyridine hydrochloride, phenazopyridine hcl, urodine, pyridium, pyripyridium, bisteril, diridone, mallophene, phenazodine, pyrazodine
InChI Key QQBPIHBUCMDKFG-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)N=NC2=C(N=C(C=C2)N)N.Cl
IUPAC Name 3-phenyldiazenylpyridine-2,6-diamine;hydrochloride
Molecular Weight (g/mol) 249.7
PubChem CID 8691
ChEBI CHEBI:71419
Formula Weight 249.7
Percent Purity 99%
Grade Analytical
Physical Form Liquid
Chemical Name or Material Phenazopyridine hydrochloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenazopyridine hydrochloride, analytical standard
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.