missing translation for 'onlineSavingsMsg'
Learn More

Phenazine Methosulfate, TRC

Product Code. 30414424
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414424 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414424 Supplier Toronto Research Chemicals Supplier No. TRCP295385500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenazine Methosulfate
CAS 299-11-6
Molecular Formula C14H14N2O4S
Quantity 500 mg
Synonym 5-N-Methylphenazonium Methosulfate, 9-Methylphenazinium Methyl Sulfate, N-Methylphenazine Methosulfate, N-Methylphenazinium Methosulfate, N-Methylphenazinium Methyl Sulfate, N-Methylphenazonium Methosulfate, N-Methylphenazonium Methosulphate, NSC 34661, PMS, PMS (pharmaceutical), Phenazine Methosulfate, Phenazine Methosulphate
InChI Formula InChI=1S/C13H11N2.CH4O4S/c1-15-12-8-4-2-6-10(12)14-11-7-3-5-9-13(11)15;1-5-6(2,3)4/h2-9H,1H3;1H3,(H,2,3,4)/q+1;/p-1
SMILES COS(=O)(=O)[O-].C[n+]1c2ccccc2nc3ccccc13
IUPAC Name 5-methylphenazin-5-ium;methyl sulfate
Molecular Weight (g/mol) 306.34
Formula Weight 306.0674
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.