missing translation for 'onlineSavingsMsg'
Learn More

Phenazine methosulfate, 98+%

Product Code. p-7037879
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15423248

Brand: Thermo Scientific Alfa Aesar H56718.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 299-11-6 | C14H14N2O4S | 306.336 g/mol

It is used with ascorbic acid to determine nitric oxide reductase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used with ascorbic acid to determine nitric oxide reductase activity.

Solubility
Soluble in water. (100g/L at20°C)

Notes
Light sensitive. Incompatible with oxidizing agents. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299-11-6
Molecular Formula C14H14N2O4S
Molecular Weight (g/mol) 306.336
MDL Number MFCD00011923
InChI Key RXGJTUSBYWCRBK-UHFFFAOYSA-M
Synonym phenazine methosulfate, 5-methylphenazin-5-ium methyl sulfate, 5-methylphenazinium methyl sulfate, n-methylphenazonium methosulfate, phenazine methosulphate, methylphenazonium methosulfate, 5-methylphenazine methylsulfate, n-methylphenazinium methosulfate, n-methylphenazonium methosulphate, phenazinium, 5-methyl-, methyl sulfate
PubChem CID 9285
ChEBI CHEBI:8055
IUPAC Name 5-methylphenazin-5-ium;methyl sulfate
SMILES C[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.COS(=O)(=O)[O-]

Specifications

Quantity 5 g
Beilstein 3898869
Sensitivity Light sensitive
Solubility Information Soluble in water. (100g/L at20°C)
Formula Weight 306.34
Percent Purity ≥98%
Chemical Name or Material Phenazine methosulfate
compliance-icons
Product Identifier
  • Phenazine methosulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Phenazine methosulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.