missing translation for 'onlineSavingsMsg'
Learn More

Phenazine methosulfate, 98+%

Product Code. 15433248
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433248 25 g 25g
15423248 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15433248 Supplier Thermo Scientific Alfa Aesar Supplier No. H56718.14
  • ONLINE EXCLUSIVE PRICE
    £392.00
    £352.65 / 25g
    Save £39.35
    10% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 299-11-6 | C14H14N2O4S | 306.336 g/mol

It is used with ascorbic acid to determine nitric oxide reductase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used with ascorbic acid to determine nitric oxide reductase activity.

Solubility
Soluble in water. (100g/L at20°C)

Notes
Light sensitive. Incompatible with oxidizing agents. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299-11-6
Molecular Formula C14H14N2O4S
Molecular Weight (g/mol) 306.336
MDL Number MFCD00011923
InChI Key RXGJTUSBYWCRBK-UHFFFAOYSA-M
Synonym phenazine methosulfate, 5-methylphenazin-5-ium methyl sulfate, 5-methylphenazinium methyl sulfate, n-methylphenazonium methosulfate, phenazine methosulphate, methylphenazonium methosulfate, 5-methylphenazine methylsulfate, n-methylphenazinium methosulfate, n-methylphenazonium methosulphate, phenazinium, 5-methyl-, methyl sulfate
PubChem CID 9285
ChEBI CHEBI:8055
IUPAC Name 5-methylphenazin-5-ium;methyl sulfate
SMILES C[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.COS(=O)(=O)[O-]

Specifications

Quantity 25 g
Beilstein 3898869
Sensitivity Light sensitive
Solubility Information Soluble in water. (100g/L at20°C)
Formula Weight 306.34
Percent Purity ≥98%
Chemical Name or Material Phenazine methosulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.