missing translation for 'onlineSavingsMsg'
Learn More

Phenazine methosulfate, 98%

Product Code. 10061310 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10061310 25 g Glass bottle 25g
10626332 1 g Glass bottle 1g
10785581 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10061310 Supplier Thermo Scientific Acros Supplier No. 130160250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 299-11-6 | C13H11N2·CH3O4S | 306.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299-11-6
Molecular Formula C13H11N2·CH3O4S
Molecular Weight (g/mol) 306.33
MDL Number MFCD00011923
InChI Key RXGJTUSBYWCRBK-UHFFFAOYSA-M
Synonym phenazine methosulfate, 5-methylphenazin-5-ium methyl sulfate, 5-methylphenazinium methyl sulfate, n-methylphenazonium methosulfate, phenazine methosulphate, methylphenazonium methosulfate, 5-methylphenazine methylsulfate, n-methylphenazinium methosulfate, n-methylphenazonium methosulphate, phenazinium, 5-methyl-, methyl sulfate
PubChem CID 9285
ChEBI CHEBI:8055
IUPAC Name 5-methylphenazin-5-ium;methyl sulfate
SMILES C[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.COS(=O)(=O)[O-]

Specifications

Melting Point 158°C to 160°C
Color Brown to Yellow or Khaki
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 6182
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol and ethanol,insoluble in benzene and diethylether
Formula Weight 306.33
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Phenazine methosulfate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenazine methosulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.