missing translation for 'onlineSavingsMsg'
Learn More

Phenazine Ethosulfate, TRC

Product Code. 30420737
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420737 500 mg -
30425610 250 mg 250mg
30425864 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420737 Supplier Toronto Research Chemicals Supplier No. TRCP295378500MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10510-77-7
Molecular Formula C14H13N2 .C2H5O4S
Molecular Weight (g/mol) 334.4
Synonym N-Ethylphenazinium Ethyl Sulfate, N-Ethylphenazonium Ethyl Sulfate, NSC 402863
IUPAC Name (E)-3-(2,5-dimethylanilino)-1-phenylprop-2-en-1-one
SMILES Cc1ccc(C)c(N\C=C\C(=O)c2ccccc2)c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C17H17NO/c1-13-8-9-14(2)16(12-13)18-11-10-17(19)15-6-4-3-5-7-15/h3-12,18H,1-2H3/b11-10+
Formula Weight 251.131
Chemical Name or Material Phenazine Ethosulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.